Skip to Content
Merck
All Photos(2)

Key Documents

MAB1217-I

Sigma-Aldrich

Anti-Fmc-7 (B-Cell Lymphocyte Marker), clone Fmc-7 Antibody

clone Fmc-7, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
€432.00

€432.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
100 TESTS
€432.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€432.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Fmc-7, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgMκ

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MS4A1(931)

Related Categories

General description

B cells are lymphocytes that play a large role in the humoral immune response as opposed to the cell-mediated immune response that is governed by T cells. The principal function of B cells is to make antibodies against soluble antigens. B cells are an essential component of the adaptive immune system. FMC-7 B-Cell Lymphocyte Marker is a 105 kDa B cell restricted antigen which is expressed on about 50% of adult human peripheral blood B cells. Upon in vivo B cell activation FMC7 expression initially increases. B cells involved in antibody secretion have lost the FMC7 determinant. The monoclonal antibody FMC7 delineates a subpopulation of B lymphocytes in normal blood. Expression of the antigen recognized by FMC7 appears to be maturation-linked, and it serves to distinguish different types of B cell leukemia.

Immunogen

HRIK [human B-lymphoblastoid cell] corresponding to human Fmc-7 (B-Cell Lymphocyte Marker).

Application

Flow Cytometry Analysis: 4.0 µg of this antibody detected Fmc-7 (B-Cell Lymphocyte Marker) in peripheral blood mononuclear cells.
This Anti-Fmc-7 (B-Cell Lymphocyte Marker) antibody is validated for use in WB, FC for the detection of Fmc-7 (B-Cell Lymphocyte Marker).

Quality

Evaluated by Western Blotting in THP-1 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected Fmc-7 (B-Cell Lymphocyte Marker) in 10 µg of THP-1 cell lysate.

Target description

~105 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing 0.1M Tris, 10mM Glycine, 0.5M NaCl, and 0.1% Sodium Azide.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service