Skip to Content
Merck
All Photos(1)

Documents

MAB1152

Sigma-Aldrich

Anti-Interferon-γ Antibody, clone RMMG-1

clone RMMG-1, Chemicon®, from rat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RMMG-1, monoclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
neutralization: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... IFNG(3458)

Specificity

Reacts with mouse Interferon gamma (IFNgamma).

Immunogen

Mouse Interferon gamma

Application

Anti-Interferon-γ Antibody, clone RMMG-1 is an antibody against Interferon-γ for use in ELISA, IP, WB, IH, NEUT.
Neutralizes mouse IFNgamma; binds to mouse IFNgamma with high affinity (KA= 10E9 M-1)

Western blot (1:500)

Immunoprecipitation

Immunohistochemistry

ELISA

Optimal working dilutions must be determined by end user.
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors

Physical form

Format: Purified
Purified immunoglobulin. Liquid in PBS containing 0.1% bovine serum albumin.

Storage and Stability

Maintain frozen at -20°C or below for retention of full activity, for up to 12 months. When thawing, the contents of the tube should be apportioned in separate tubes so that freezing and thawing is kept to a minimum.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alma Zernecke et al.
Blood, 107(11), 4240-4243 (2006-02-10)
The chemokine RANTES has been implicated in neointimal hyperplasia after arterial injury. We analyzed the differential role of the RANTES receptors CCR1 and CCR5 by genetic deletion in apolipoprotein E-deficient mice. Deficiency in CCR5 significantly reduced neointimal area after arterial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service