Skip to Content
Merck
All Photos(1)

Key Documents

MAB10764

Sigma-Aldrich

Anti-SBP-tag Antibody, clone 20 | MAB10764

clone 20, 1.0 mg/mL, from mouse

Synonym(s):

streptavidin binding peptide

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€544.00

€544.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€544.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€544.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

20, monoclonal

species reactivity

human

concentration

1.0 mg/mL

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

shipped in

wet ice

target post-translational modification

unmodified

General description

Streptavidin Binding Peptide tag is a useful and versatile peptide that can be inserted into expression vectors for easy detection of recombinant proteins and purification. The sequence for SBP-tag is 38 amino acids long having a nanomolar scale binding affinity for streptavidin with an equilibrium constant of 2.5 nM.

Specificity

Demonstrated to react with cells expressing the SBP tag.
This antibody recognizes SBP tag.

Immunogen

Epitope: Unknown
Linear peptide corresponding to SBP tag.

Application

Immunofluorescence Analysis: A previous lot was used by an independent laboratory in IF. (Hudson, D., et al. (2008). MBC. 19(7):3070-3079.)
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags

Quality

Evaluated by Western Blot in pTrAP-GFP transfected Hek293 lysate.

Western Blot Analysis: 0.1 ng/mL of this antibody detected SBP-tag on 10 µg of pTrAP-GFP transfected Hek293 lysate.

Target description

The protein expresss is pTrAP-GFP plasmids containing a Histidine tag, S-tag, streptavidin-binding pepide (SBP) tag, and GFP. The protein expressing is ~37 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
pTrAP-GFP transfected Hek293 lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashley J Evans et al.
Cell reports, 19(12), 2613-2626 (2017-06-22)
Ionotropic glutamate receptor (iGluR) trafficking and function underpin excitatory synaptic transmission and plasticity and shape neuronal networks. It is well established that the transcription, translation, and endocytosis/recycling of iGluRs are all regulated by neuronal activity, but much less is known
Gaelle Boncompain et al.
Methods in molecular biology (Clifton, N.J.), 1174, 211-223 (2014-06-21)
In mammalian cells, secretory proteins are transported to their destination compartment via the secretory pathway. Cargos start their journey in the endoplasmic reticulum and then reach the Golgi complex where they are processed and sorted to be delivered to their
Christopher M Ziegler et al.
Pathogens (Basel, Switzerland), 7(4) (2018-12-14)
Lassa mammarenavirus (LASV) is an enveloped RNA virus that can cause Lassa fever, an acute hemorrhagic fever syndrome associated with significant morbidity and high rates of fatality in endemic regions of western Africa. The arenavirus matrix protein Z has several
Laura Berneking et al.
Bio-protocol, 9(12), e3277-e3277 (2019-06-20)
Identification of protein-protein interactions of bacterial effectors and cellular targets during infection is at the core to understand how bacteria manipulate the infected host to overcome the immune response. Potential interacting proteins might be identified by genetic methods, i.e., two
Miaomiao Li et al.
Nature communications, 14(1), 2600-2600 (2023-05-06)
Many eukaryotic transcription factors (TF) form homodimer or heterodimer complexes to regulate gene expression. Dimerization of BASIC LEUCINE ZIPPER (bZIP) TFs are critical for their functions, but the molecular mechanism underlying the DNA binding and functional specificity of homo- versus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service