Skip to Content
Merck
All Photos(1)

Key Documents

MAB10207-I

Sigma-Aldrich

Anti-Fusion Glycoprotein of HPIV 3 Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€153.00
100 μL
€383.00

€153.00


Estimated to ship on17 April 2025


Request a Bulk Order

Select a Size

Change View
25 μL
€153.00
100 μL
€383.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€153.00


Estimated to ship on17 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

9-4-3, monoclonal

mol wt

calculated mol wt 60.06 kDa

species reactivity

virus

packaging

antibody small pack of 100 μL

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

Fusion glycoprotein F0 (UniProt: P06828) is encoded by the F gene (Gene ID: 2828653) by the Human parainfluenza 3 virus (HPIV-3). HPIVs are antigenically divided into types 1-4 and type 1 and 3 are reported to cause most lower respiratory infections. HPIVs are enveloped and of medium size (150 to 250 nm), and their RNA genome is in the negative sense. HPIV3 displays three conformational states: pre-fusion native state, pre-hairpin intermediate state, and post-fusion hairpin state. During viral and plasma cell membrane fusion, the heptad repeat (HR) regions assume a trimer-of-hairpins structure, positioning the fusion peptide near the C-terminal region of the ectodomain. The formation of this structure drives apposition and subsequent fusion of viral and plasma cell membranes. This fusion is reported to be pH independent and occurs directly at the outer cell membrane. Fusion glycoprotein F0 is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-18), which is subsequently cleaved off to generate the mature form. This inactive precursor F0 is glycosylated and proteolytically cleaved into F1 (aa 110-539) and F2 (19-109) to be functionally active. The cleavage is mediated by cellular proteases during the transport and maturation of the polypeptide. (Ref.: Hendrickson, KJ. (2003). Clin. Microbiol. Rev. 16(2); 242-264).

Specificity

Clone 9-4-3 is mouse monoclonal antibody that detects Fusion glycoprotein of Parainfluenza 3 virus.

Immunogen

Fusion (F) glycoprotein of human parainfluenza type 3 virus.

Application

Quality Control Testing

Evaluated by Immunocytochemistry in LLC-ML2 cells infected with HPIV-3.

Immunocytochemistry Analysis: A 1:250 dilution of this antibody detected Fusion Glycoprotein of HPIV 3 in LLC-ML2 cells infected with HPIV-3.

Tested Applications

Western Blotting Analysis: A representative lot detected Fusion Glycoprotein of HPIV-3 in Western Blotting applications (Xie, W., et. al. (2015). PLoS One. 10(8):e0136474).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Fusion Glycoprotein of HPIV 3, clone 9-4-3, Cat. No. MAB10207-I, is a mouse monoclonal antibody that detects human parainfluenza type 3 virus and is tested for use in Immunocytochemistry and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service