Skip to Content
Merck
All Photos(1)

Key Documents

MAB10139

Sigma-Aldrich

Anti-LIM Domain Only 2 Antibody, clone 2E6

culture supernatant, clone 2E6, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

2E6, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LMO2(4005)

General description

LIM Domain only 2 (LMO2) encodes a cysteine-rich, two LIM-domain protein that is required for yolk sac erythropoiesis. The LMO2 protein has a central and crucial role in hematopoietic development and is highly conserved. The LMO2 transcription start site is located approximately 25 kb downstream from the 11p13 T-cell translocation cluster (11p13 ttc), where a number T-cell acute lymphoblastic leukemia-specific translocations occur.

Specificity

Human

Immunogen

Recombinant human LMO2

Application

Detect LIM Domain Only 2 using this Anti-LIM Domain Only 2 Antibody, clone 2E6 validated for use in WB.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Routinely evaluated by immunoblot.

Target description

18kDa

Physical form

Liquid containing 0.1% sodium azide.

Storage and Stability

1 year at 4°C from date of shipment

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service