Skip to Content
Merck
All Photos(1)

Key Documents

L30DET6S1

Millipore

Millistak+® Lenticular Disc Filter

DE30, cellulose, filtration area 3.47 m2, hydrophilic

Synonym(s):

Millistak+ Lenticular Disc Filters, DE30 media series, 16 in. diameter, 16-cell, silicone seal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806
eCl@ss:
32031690
Pricing and availability is not currently available.

material

cellulose
filter aid (inorganic)
polypropylene
silicone seal

Quality Level

description

For clarification of serum, plasma, vaccines, cell culture or other fluids.

feature

hydrophilic

manufacturer/tradename

Millistak+®

parameter

0.03 bar max. differential pressure (0.5 psid) (Reverse)
0.2 bar max. inlet pressure (3 psi) at 123 °C
0.21 bar max. differential pressure (3 psid) at 123 °C (Forward)
1 bar max. inlet pressure (15 psi) at 80 °C
1.03 bar max. differential pressure (15 psid) at 80 °C (Forward)
2.07 bar max. differential pressure (30 psid) at 25 °C (Forward)
2.1 bar max. inlet pressure (30 psi) at 25 °C

H

27.6 cm (10.88 in.)

diam.

40.64 cm (16 in.)

filtration area

3.47 m2

impurities

<0.25 EU/mL bacterial endotoxins (LAL test, Aqueous extraction)

total organic carbon (TOC) residue

753-1567 ppb

input

sample type liquid

conductivity

9-12 μmho per cm at 25 °C (product water)

filter grade

DE30

Looking for similar products? Visit Product Comparison Guide

General description

Device Configuration: Stack Disk
Stack Design: 16 in. diameter, 16-cell

Application

For clarification of serum, plasma, vaccines, cell culture or other fluids.

Specifications

Filter Grade: DE30

Preparation Note

Sanitization Protocol
May be hot water sanitized for 20 cycles using purified water @ 80 °C for 30 minutes
Sterilization Method
May be steam sterilized for 3 steam-in-place (SIP) cycles of 30 minutes @ 123 °C

Analysis Note

Product Water Conductivity at 25°C: after a pure water flush of 5 liters per ft2 (50 L/m2) of media surface area per autoclaved (1 cycle of 30 minutes at 123 °C) 12" 16-stack device
The effluent of an autoclaved (1 cycle of 30 minutes @ 123 °C) stacked device is negative after a WFI flush of 5 liters per ft2 (50 L/m2) of media surface area
Total Organic Carbon (TOC) Residue: ppb post autoclave (1 cycle of 30 minutes at 123 °C) and pure water flush of 5 liters per ft2 (50 L/m2) of media surface area

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service