Skip to Content
Merck
All Photos(3)

Key Documents

KWEFHAW10B

Millipore

EZ-Fit® Family Kit

pkg of 48 units (bulk packaging with protective bag)

Synonym(s):

EZ-Fit® Family Starting Kit, EZ-Fit® Filtration system for bioburden testing, Filtration Kit, Filtration Unit

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€2,950.00

€2,950.00


Estimated to ship on22 May 2025


Request a Demo

Select a Size

Change View
1 UNIT
€2,950.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

€2,950.00


Estimated to ship on22 May 2025


Request a Demo

material

mixed cellulose esters (MCE) membrane
polystyrene cover
stainless steel (Manifold)
styrene-butadiene funnel(s)
white filter

Agency

ISO 7704

sterility

electron beam irradiated

Sterilization Compatibility

autoclavable compatible

feature

ready-to-use

packaging

pkg of 48 units (bulk packaging with protective bag)

manufacturer/tradename

EZ-Fit®

parameter

45 °C temperature (for samples)

D

47 mm

funnel capacity

100 mL

volume

100 mL

pore size

0.45 μm pore size

suitability

suitable for (suitable for solid and liquid cultivation)

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with EZ-Fit® System

shipped in

ambient

General description

EZ-Fit® Manifold Starting Kit is designed in a way to support laboratory filtration test methods. The kit contains EZ-Fit® Manifold and 4 boxes of EZ-Fit® 100ml Funnels making it the perfect kit to start sample testing through filtration.
The EZ-Fit® Filtration device is designed to fit onto EZ-Fit® Manifold, which features specific EZ-Fit® Filtration Heads.
The state-of-the-art design is optimized to secure laboratory workflow, avoid cross contamination and save testing time thereby producing reliable microbiological results.
After the filtration process is completed, the membrane can be easily transferred onto an agar plate for microbial growth.

Application

EZ-Fit® Manifold Starting Kit can be used to carry out bioburden testing of liquid samples like water, process samples or final products.

Packaging

EFHAW100B x4 - 192 units, Bulk Packaging
EZFITBASE3 x1
EZFITMVHE3 x1
EZSTREAM1 x1

Components

Contains 4 boxes of EZ-Fit® 100 mL Funnels with bulk packaging, EZ-Fit® 3-head Manifold with filtration heads, EZ-STREAM® pump

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
EZ-STREAM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service