Skip to Content
Merck
All Photos(1)

Key Documents

JM-10L-BK

Millipore

Intermediates for Further Manufacturing Use (FFMU) - ABO Specificities

Anti-B from the clone LB-2, Murine IgM, Intermediate for further manufacturing use

Synonym(s):

Anti-B (clone LB-2), FFMU

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116102
NACRES:
NA.41

Note: For further information on price, availability or to place an order, please contact us

biological source

mouse

Quality Level

antibody form

unpurified

clone

LB-2, monoclonal

form

liquid

packaging

pack of 10 L

concentration

3-15 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

6.7-7.7

isotype

IgM

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Standardized, concentrated cell culture supernatant which has been diafiltered with phosphate buffered saline.

For the formulation of Anti-B or Anti-A+B blood typing reagents or devices.

This antibody reacts strongly with all common forms of the B antigen. It has the benefit of not agglutinating unmodified or enzyme treated A or O red cells, or A<sub>1</sub> red cells with the ‘Acquired B′ antigen.

Contains 0.1% (w/v) Sodium Azide; Shelf Life: 24 months from date of filtration.

Licensed by the US FDA under a shared manufacturing agreement.

Application

Antibody intermediate for further manufacturing use.
Production of an in vitro blood typing product or device.

Legal Information

Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Partially processed reagent for further manufacture of non-injectable products only. The absence of all viral antigens has not been determined. No US Standard of Potency. Do not use if turbid. Contains 0.1%<nbsp/>(w/v) sodium azide.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service