Skip to Content
Merck
All Photos(1)

Key Documents

HTS098RTA

Sigma-Aldrich

Ready-to-Assay NMU2 Neuromedin U Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.84

product name

Ready-to-Assay NMU2 Neuromedin U Receptor Frozen Cells, Human NMU2 GPCR frozen cells for Calcium Flux FLIPR Assays.

biological source

human

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

NCBI accession no.

detection method

fluorometric

shipped in

dry ice

General description

Millipore’s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

Neuromedin U (NmU) is a peptide which regulates peripheral functions such as smooth muscle contraction and blood pressure, and CNS functions including nociception and feeding activity (Brighton et al., 2004a). Two GPCRs, NMU1 and NMU2, mediate the contractile effects of neuromedin U by activation of both Gq and Gi (Brighton et al., 2004b). Compared to the wide distribution of NMU1 in peripheral tissue, expression of NMU2 receptor is limited to areas of the brain, such as the paraventricular nucleus, along the wall of the third ventricle in the hypothalamus, the CA1 region of the hippocampus, and the spinal cord (Howard et al. 2000). Recent study has shown that mice deficient in NMU2 but not NMU1 receptor had impaired nociceptive responses, suggesting that the pro-nociceptive effects of NmU in mice appear to be mediated through NMU2 (Zeng et al., 2006; Torres et al. 2007). Millipore’s cloned human NMU2-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant NMU2 expression on the cell surface for functional detection via the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at NMU2.

Cell Line Description

Chem-1
GPCR Cell Lines

Application

Human NMU2 GPCR frozen cells for Calcium Flux FLIPR Assays.

Biochem/physiol Actions

GPCR Class: A
Protein Target: NMU2
Target Sub-Family: Neuromedin U

Components

Pack contains 1 vial of mycoplasma-free cells, 1 ml per vial. Fifty (50) mL of Media Component.

Disclaimer

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service