Skip to Content
Merck
All Photos(1)

Key Documents

HTS080RTA

Sigma-Aldrich

Ready-to-Assay NK1 Tachykinin Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76

Product Name

Ready-to-Assay NK1 Tachykinin Receptor Frozen Cells,

biological source

human

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

NCBI accession no.

detection method

fluorometric

General description

Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

The tachykinin peptide family in mammals comprises three peptides, substance P, neurokinin A and neurokinin B, which bind to the tachykinin receptor family of GPCRs, NK1, NK2 and NK3 (Severini et al., 2002). Tachykinins have prominent activity in the GI system, in which they stimulate intestinal contraction and salivation. These effects are mediated by NK1 and NK2, and an antagonist of NK1, aprepitant, is used for treatment of chemotherapy-induced emesis (Rupniak and Kramer, 1999). The NK1 tachykinin receptor is expressed in brain, and is thought to be involved in depression and nociception (Saria, 1999). Although NK1 appears to promote nociception in animal models, the significance of this activity is controversial, as antagonists of NK1 have not proven efficacious in pain relief in humans (Hill, 2000). Millipore′s cloned human NK1-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant NK1 expression on the cell surface and contains high levels of the promiscuous G protein Gα15 to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for antagonists of interactions between NK1 and its ligands.

Cell Line Description

  • GPCR Cell Lines
  • Host cells: Chem-1

Application

Human NK1 GPCR frozen cells for Calcium Flux FLIPR Assays.

Biochem/physiol Actions

  • GPCR Class: A
  • Protein Target: NK1
  • Target Sub-Family: Tackykinin/ neurokinin

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial.
Fifty (50) mL of Media Component.

Disclaimer


This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service