HS10/4XS-N
NovAseptic® Mixer HS T10 compatible Drive Unit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
parameter
max. pressure at 25 °C (atmospheric pressure)
-15-40 °C max. temp. (5-104 °F)
Quality Level
General description
NovAseptic® mixers offer a complete solution for mixing throughout the
process line and are designed for a wide variety of mixing applications in the
pharmaceutical and biotechnology industries.
process line and are designed for a wide variety of mixing applications in the
pharmaceutical and biotechnology industries.
Specificity
General Motor Data:Motor Type: Nord 63LP/2 IE3 Frequency: 50 Hz, Speed:2770 rpmDrive Unit Frame: IEC 63 B14Design Speed: 350-5200rpm
Application
NovAseptic® mixers offer a complete solution for mixing throughout the
process line and are designed for a wide variety of mixing applications in the
pharmaceutical and biotechnology industries.
process line and are designed for a wide variety of mixing applications in the
pharmaceutical and biotechnology industries.
Components
The drive unit is packaged in a closed box.
Legal Information
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service