The BD P800 tubes contain inhibitors for GLP-1 active, Amylin, and Glucagon, but not for Active Ghrelin. If you also want to include active Ghrelin, you will need to add AEBSF.
HMH3-34K
MILLIPLEX® HUMAN METABOLIC HORMONE PANEL V3
Synonym(s):
human metabolic hormone immunoassay panel, luminex human metabolic hormone multiplex assay, millipore human metabolism multiplex kit
About This Item
Recommended Products
species reactivity
human
Quality Level
manufacturer/tradename
Milliplex®
technique(s)
multiplexing: suitable
detection method
fluorometric (Luminex® xMAP®)
storage temp.
2-8°C
General description
Specificity
Application
Features and Benefits
Packaging
Storage and Stability
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Simplify protein biomarker discovery with the MILLIPLEX® PLEXpedition 115-Plex Multiplex Screening Panel. Effortlessly screen cytokines and other markers with Luminex® technology.
Multiplex immunoassays, such as MILLIPLEX® multiplex metabolic assays, are critical in metabolic syndrome research because they provide a full picture of the different conditions related to it, like diabetes and obesity, as well as save time and sample volume.
Discover the benefits of MILLIPLEX® multiplex assays, based on Luminex® xMAP® multiplex assay technology, that provide consistent, high-quality results and see how these multiplex biomarker Luminex® assays are being used to advance research.
-
Would the BD P800 sample collection tubes be suitable for use in the Milliplex Human Metabolic Hormone Panel?
1 answer-
Helpful?
-
-
Could you provide me with the differences between the Human Metabolic Hormone Expanded Panel (HMHEMAG-34K) and the new Human Metabolic Hormone Panel V3 (HMH3-34K)?
1 answer-
The following changes have been implemented:
- Secretin has been added
- Assay Buffer has been changed to LE-ABGLP3 to enhance Insulin and Glucagon Detection
- The antibody pair for Insulin, IL-6, MCP-1, PP, and TNF-a has been replacedHelpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service