Skip to Content
Merck
All Photos(1)

Key Documents

HAWP047A0

Millipore

AO Pak Filters 0.45µm 47mm white plain

Microbiological analysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

mixed cellulose esters (MCE)
plain filter
white filter

sterility

non-sterile

feature

autoclavable
hydrophilic

packaging

box of 10 envelopes containing 10 pak

filter diam.

47 mm

pore size

0.45 μm pore size

input

sample(s) (environmental)
sample type beverage(s)
sample type cosmetics
sample type food(s)
sample type sport drink
sample type beer
sample type wine
sample type cider
sample type soft drink
sample type bottled water

application(s)

bioburden testing
clinical
cosmetics
environmental
environmental
food and beverages
industrial qc

shipped in

ambient

General description

AO Pak filters - 10 resealable envelopes containing 10 filters and 10 pads, requires autoclaving.

Application

Microbiological analysis

Packaging

Autoclave pack: 10 resealable envelopes containing 10 filters and 10 pads, requires autoclaving

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service