GM5/120EPDC
NovAseptic® GMP 500 Mixing Head, Male BearingGMP (UNCD Coated)
About This Item
material
316L stainless steel plate
EPDM rubber O-ring
ceramic bearing (Male Bearing UNCD coating)
Quality Level
parameter
5-135 °C temp. range (41-275 °C)
roughness average (RA)
≤0.38 μm (Surface Treatment: Electropolished and UNCD coated)
pH range
1-14
General description
Packaging
Components
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service