Skip to Content
Merck
All Photos(1)

Key Documents

FX0235

Supelco

Ferric Sulfate, Hydrate

GR

Synonym(s):

Ferric Sulfate, Hydrate, Iron(III) Sulfate, Hydrate, Ferric Persulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe2(SO4)3 · xH2O
CAS Number:
MDL number:
UNSPSC Code:
12352300
EC Index Number:
233-072-9
Assay:
≥72.0% (Fe2(SO4)3 basis)
form:
powder

Quality Level

Assay

≥72.0% (Fe2(SO4)3 basis)

form

powder

impurities

≤0.02% Insoluble matter
≤0.1% Substances not precipitated by ammonium hydroxide

color

slight yellow

pH

1.5 (20 °C, 50 g/L in H2O)

mp

480 °C (decomposition)

anion traces

chloride (Cl-): ≤0.01%
nitrate (NO3-): ≤0.02%

cation traces

Cu: ≤0.005%
Fe2+: ≤0.02%
Zn: ≤0.02%

shipped in

ambient

storage temp.

room temp

InChI

1S/2Fe.3H2O4S.H2O/c;;3*1-5(2,3)4;/h;;3*(H2,1,2,3,4);1H2/q2*+3;;;;/p-6

InChI key

VXWSFRMTBJZULV-UHFFFAOYSA-H

Application

GR

Analysis Note

Assay [as Fe2(SO4)3]: 72.0% min
Chloride (Cl): 0.01% max
Color: Slight yellow
Copper (Cu): 0.005% max
Ferrous iron (Fe++): 0.02% max
Insoluble matter: 0.02% max
Nitrate (NO3): 0.02% max
Substances not precipitated by ammonium hydroxide: 0.10% max
Zinc (Zn): 0.02% max

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service