FR180380000
Estapor® Magnetic Microspheres
M1-030/40 Mag COOH PS 10%,Magnetic microspheres 300nm, ChemiLuminsecence Immunoassay, Lateral Flow Immunoassay
Synonym(s):
Magnetic Beads, Magnetic Particles
About This Item
Recommended Products
material
high-density polyethylene packaging (bottle)
polystyrene beads
Quality Level
composition
Iron Content, 40.0-60.0%
manufacturer/tradename
Estapor®
packaging
bottle of 10ml-10L
concentration
10% solids
technique(s)
enzyme immunoassay: suitable (ChemiLuminsecence Immunoassay-CLIA)
color
brown bead
matrix
polymeric polystyrene/divinylbenzene
bead diameter
0.251—0.400 μm
capacity
100—250 μeq/g surface charge density
functional group
carboxylic acid (COOH)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
the microsphere of choice for many CLIA and
bio-separation based diagnostic manufacturers. Used globally in millions of assays made by internationally known brands, our microspheres are adapted to both manual and automated platforms, are highly characterized, customizable and designed with key features to ideally meet the needs of IVD manufacturers.
Application
Thanks to their small diameter, these microspheres can also be used as reporter particles in qualitative or quantitative Later Flow ImmunoAssays
Features and Benefits
- High magnetic susceptibility
- Low non-specific binding
- Good colloidal stability
- Fast magnetic response time and dispersion
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Estapor® Microspheres, for polymeric supports in diagnostics, life sciences, biotechnology, & environmental applications, offers 100+ microsphere types.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service