Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB230P

Sigma-Aldrich

Milli-Mark® Mouse IgG1-k, clone MOPC-21, PE conjugate

This Milli-Mark Anti-mouse IgG1-PE Antibody, clone MOPC-21 is validated for use in FC for the detection of mouse IgG1.

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
€390.00

€390.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 TESTS
€390.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€390.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

PE

antibody form

purified immunoglobulin

clone

MOPC-21, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

shipped in

wet ice

target post-translational modification

unmodified

General description

The MOPC-21 tumor line that produces mouse IgG1,k, is a mineral oil induced plasmacytoma. The product is originated and carried intraperitoneally in BALB/c mice. Identity and purity of the immunoglobulin is established by immunoelectrophoresis (IEP) and Ouchterlony Double Diffusion (ODD), prior to conjugation. Electrophoresis of the purified immunoglobulin, followed by diffusion versus anti-mouse whole serum, antimouse IgG1 and anti-mouse kappa chain, results in single arcs of precipitation. By ODD the purified immunoglobulin preparation is non-reactive with antisera to mouse lambda light chain, mouse IgA, IgM, IgG2a, IgG2b, and IgG3. The specificity of staining by monoclonal antibodies to human CD antigens should be verified by establishing the amount of non-specific binding to the target cell population. It is recommended that a non-reactive immunoglobulin of the same isotype, concentration and F/P molar ratio be included as a negative control for each monoclonal antibody reagent used in flow cytometry or other immunoassays.

Specificity

This mouse IgG1 isotype control antibody is not reactive with any known antigen.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Isotype Control Antibodies

Quality

Evaluated for use as flow cytometry isotype control using Jurkat Cells.

Physical form

Immunoelectrophoresis
Purified mouse monoclonal IgG1κ conjugated to PE in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robin F Chan et al.
Nucleic acids research, 45(11), e97-e97 (2017-03-24)
Methylome-wide association studies are typically performed using microarray technologies that only assay a very small fraction of the CG methylome and entirely miss two forms of methylation that are common in brain and likely of particular relevance for neuroscience and
Charles Gullo et al.
PloS one, 5(5), e10845-e10845 (2010-06-04)
Multiple myeloma (MM) is a malignancy of terminally-differentiated plasma cells, and the second most prevalent blood cancer. At present there is no cure for MM, and the average prognosis is only three to five years. Current treatments such as chemotherapy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service