Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB195F

Sigma-Aldrich

Milli-Mark® Anti-CD41-FITC Antibody, clone 5B12

clone 5B12, Milli-Mark®, from mouse

Synonym(s):

Integrin alpha-IIb, Platelet membrane glycoprotein IIb, GPalpha IIb, GPIIb

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
€390.00

€390.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 TESTS
€390.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

€390.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5B12, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA2B(3674)

General description

CD41 is the α-subunit of the CD41/CD61 complex (GPIIb-IIIa), a calcium-dependent, non-covalently associated heterodimer. CD41 is a 135 kDa molecule, consisting of 2 peptides: a 120 kDa α chain and a 23 kDa β chain. The α chain and the β chain are linked by a single disulphide bond. The α chain contains four calcium-binding sites and is entirely extracellular, while the β-chain has extracellular, transmembrane, and cytoplasmic domains (Sun, 1997). The CD41/CD61 complex appears early in megakaryocyte maturation (Vinci, 1984). Anti-CD41, 5B12, was included in the Fifth International Workshop and Conference on Human Leukocyte Differentiation Antigens and studies by a number of laboratories confirmed its reactivity with CD41 (Honda, 1995).

Specificity

Antibody recognizes Human CD41 antigen.

Immunogen

Normal human platelets

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Milli-Mark Anti-CD41-FITC Antibody, clone 5B12 is validated for use in FC for the detection of CD41.

Quality

Evaluated by flow cytometry using TF-1 cells

Target description

135 kDa (Sun, 1997)

Physical form

Protein A purified
Purified Mouse monoclonal IgG1 conjugated to FITC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Storage and Stability

Maintain refrigerated at 2-8°C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
TF-1 cells

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesca Patella et al.
Research and practice in thrombosis and haemostasis, 5(8), e12626-e12626 (2021-12-23)
Thrombotic thrombocytopenic purpura (TTP), caused by a genetic or autoimmune-driven lack of ADAMTS-13 activity, leads to high levels of the ultra-large von Willebrand factor (VWF) multimers produced by endothelial cells, causing excess platelet recruitment into forming thrombi, often with mortal
Francesca Patella et al.
Journal of cell science, 133(14) (2020-06-25)
The haemostatic protein von Willebrand factor (VWF) exists in plasma and subendothelial pools. The plasma pools are secreted from endothelial storage granules, Weibel-Palade bodies (WPBs), by basal secretion with a contribution from agonist-stimulated secretion, and the subendothelial pool is secreted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service