Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB103AP

Sigma-Aldrich

Milli-Mark Anti-Ki67 Antibody, clone Ki-S5 APC conjugate

clone Ki-S5, Milli-Mark®, from mouse

Synonym(s):

MKI67

KIA

Ki-67

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
€381.00

€381.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 TESTS
€381.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€381.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

Allophycocyanin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Ki-S5, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MKI67(4288)

Related Categories

General description

Ki67 antigen is the prototypic cell cycle related nuclear protein, expressed by proliferating cells in all phases of the active cell cycle (G1, S, G2 and M phase). It is absent in resting (G0) cells. Ki67 antibodies are useful in establishing the cell growing fraction in neoplasms (immunohistochemically quantified by determining the number of Ki67 positive cells among the total number of resting cells = Ki67 index). In neoplastic tissues the prognostic value is comparable to the tritiated thymidine labelling index. The correlation between low Ki67 index and histologically low grade tumours is strong. Ki67 is routinely used as a marker of cell cycling and proliferation. This specificity of Ki-S5 antibody for proliferating cells might make it a useful tool for determination of the proliferative fraction in tumors such as Non-Hodgkin′s lymphoma (Kreipe et al., 1993), mammary carcinomas (Mauri et al., 1984), ovarian tumors (Ballin et al., 1994) and prostate cancer.

Specificity

Antibody recognizes Ki67.

Immunogen

Nuclear protein preparation from the human cell line U937.50

Application

Detect Ki67 using this Milli-Mark Anti-Ki67 Antibody, clone Ki-S5 APC conjugate validated for use in FC, IC, IH, FC, IH(P).
Research Category
Signaling
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Evaluated by Flow Cytometry with Jurkat cells.

Target description

345 & 395 kDa observed

Physical form

Protein A purified
Purified mouse monoclonal IgG1 conjugated to Allophycocyanin (APC) in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8oC in undiluted aliquots for up to 6 months from date of receipt. Protect from light.

Analysis Note

Control
Stimulated Jurkat Cells

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Henrikki Mertaniemi et al.
Biomaterials, 82, 208-220 (2016-01-15)
Upon surgery, local inflammatory reactions and postoperative infections cause complications, morbidity, and mortality. Delivery of human adipose mesenchymal stem cells (hASC) into the wounds is an efficient and safe means to reduce inflammation and promote wound healing. However, administration of
Margarita Pustovalova et al.
International journal of molecular sciences, 21(9) (2020-05-14)
Radiation therapy is one of the main methods of treating patients with non-small cell lung cancer (NSCLC). However, the resistance of tumor cells to exposure remains the main factor that limits successful therapeutic outcome. To study the molecular/cellular mechanisms of
Xiao-Tong Lin et al.
eLife, 10 (2021-11-16)
Dysregulation of tumor-relevant proteins may contribute to human hepatocellular carcinoma (HCC) tumorigenesis. FBXO45 is an E3 ubiquitin ligase that is frequently elevated expression in human HCC. However, it remains unknown whether FBXO45 is associated with hepatocarcinogenesis and how to treat

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service