Skip to Content
Merck
All Photos(2)

Documents

EZSWIRE01

Millipore

Power Connector for EZ-Stream® Vacuum Pump

Power Cable for uninterrupted filtration

Synonym(s):

Power Cable, Power Connetor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116500
NACRES:
NB.79

feature

repair kit

packaging

pkg of 1 unit

manufacturer/tradename

EZ-Stream®

application(s)

bioburden testing
food and beverages
industrial qc
pharmaceutical
water monitoring

compatibility

for use with EZ-Stream® Vacuum Pump

General description

Power Connector for EZ-Stream® Vacuum Pump is designed supply power to the vacuum pump during filtration. It is an accessory system that can be ordered easily.
This reference includes the power connector that fit in pump housing and the hexagonal nut for fastening the connector to the housing. The power connector is already wired with the electronic board connector for easy replacement without soldering.
The EZ-Stream® Vacuum Pump is ideal to be used both on work bench and laminar flow hood, where its small footprint saves laboratory space.

Application

Power Connector for EZ-Stream® Vacuum Pump is used to supply power to the vacuum pump during the filtration process.

Legal Information

EZ-STREAM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service