Skip to Content
Merck

Skip To

ES009

p-NPP Single Reagent, Yellow, Alkaline Phosphatase Substrate (soluble)

500 ml p-NPP Single Reagent, Yellow, Alkaline Phosphatase Substrate (soluble) for ELISA.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mL
Check Cart for Availability
€ 315,00

About This Item

UNSPSC Code:
41116010
NACRES:
NB.22
eCl@ss:
42029053

€ 315,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

λmax

405 nm

Application

p-Nitrophenyl phosphate (p-NPP) is the customary product utilized for the assay of alkaline phosphatase. It is usually supplied in tablet or powder form and is dissolved in an appropriate buffer immediately prior to assay. Liquid p-NPP concentrates are also available, but must also be diluted in buffer. CHEMICON, Inc. now provides p-NPP stabilized in diethanolamine (DEA) buffer and available for instant use. The DEA buffer contains Mg++ and Zn++ ions providing optimum activation of alkaline phosphatase labeled probes.

This is an excellent substrate for phosphatase based ELISA assays.

It produces a soluble end product which is yellow in color and reads at 405 nm to 410 nm. Both kinetic and end point determinations can be performed.

Physical form

Color Code: Yellow
p-NPP, a single component substrate in proprietary stabilizer.

Preparation Note

Maintain at 2-8°C for up to one year.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P7998ES001ES022
technique(s)

ELISA: suitable

technique(s)

-

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable, immunohistochemistry: suitable

form

liquid

form

liquid

form

liquid

form

liquid

manufacturer/tradename

Chemicon®

manufacturer/tradename

-

manufacturer/tradename

Chemicon®

manufacturer/tradename

Chemicon®

λmax

405 nm

λmax

-

λmax

450 nm

λmax

450 nm


pictograms

Health hazardCorrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Irrit. 2

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters