EFHAB250
EZ-Fit® filtration unit with absorbent pad
pore size 0.45 μm, mixed cellulose esters (MCE) membrane, white filter, volume (250 mL), pkg of Single packaging (EFHAB250I), pkg of Bulk packaging (EFHAB250B), suitable for (suitable for solid and liquid cultivation)
Synonym(s):
Membrane filtration device made of gridded, mixed cellulose esters membrane for bioburden testing
Select a Size
About This Item
Recommended Products
material
gridded filter
mixed cellulose esters (MCE) membrane
white filter
Quality Level
100
300
Agency
ISO 7004
sterility
electron beam irradiated
feature
ready-to-use
manufacturer/tradename
EZ-Fit®
packaging
pkg of Bulk packaging (EFHAB250B)
pkg of Single packaging (EFHAB250I)
capacity
250 mL
filter diam.
47 mm
funnel capacity
250 mL
volume
(250 mL)
pore size
0.45 μm pore size
input
sample type food(s)
sample type pharmaceutical(s)
sample type beverage(s)
suitability
suitable for (suitable for solid and liquid cultivation)
application(s)
bioburden testing
food and beverages
pharmaceutical
water monitoring
compatibility
for use with EZ-Fit® System
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The filtration units are e-beam irrated, a vented lid covers the sample throughout filtration, and a 360° rim prevents forceps from accidentally touching the membrane filtration area during membrane transfer, to protect samples from contamination while ensuring reliable microbial enumeration results.
The EZ-FIT® Azure feature makes the EZ-FIT® filtration unit even more ergonomic and convenient with transparent funnel and all-around filling level indicators for an easy filling from any angle. The innovative funnel assembly design prevents any leaks or sample by-pass of the membrane.
The blue units provide the extra flexibility of either transferring the membrane to an agar plate or converting the device into a Petri dish by adding the liquid media, after filtration.
The filtration units conform to international standards and minimize sample residue. The units can be stacked to save the work area. This disposable device is designed to fit onto the EZ-FIT® Manifold, which features specific EZ-FIT® Filtration heads.
Application
Packaging
EFHAB250B: Bulk packaging with protective bag (48 units/pack)
Legal Information
configured for
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Comparison of convenience, operating time, accuracy, costs and safety aspects of single-use funnels vs. reusable funnels for the membrane filtration method in QC microbiology.
The EZ Product Family optimizes your microbial testing workflow to deliver reliable and reproducible microbial enumeration test results. The product lineup consists of filtration devices, manifolds, calibrated vacuum sources, and rapid microbial detection platforms.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service