Skip to Content
Merck

ECCL2

MPAGE®® Electrode Core Clamp

replacement part for mPAGE® Mini Gel Tank

Synonym(s):

electrode assembly

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41105305
NACRES:
NC.01
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

mPAGE® Electrode Core Clamp, 2pk, replacement part for mPAGE® Mini Gel Tank

packaging

pkg of 2 packs

manufacturer/tradename

Millipore

technique(s)

SDS-PAGE: suitable, electrophoresis: suitable

compatibility

for use with mPAGE® Mini Gel Tank

General description

mPAGE® Electrode Core Clamp, 2pk, for use with mPAGE® Mini Gel Tank (MGT-2 and MGT-4).

The mPAGE® Mini Gel Tank is used to perform high-resolution protein separation on handcast or precast polyacrylamide gels. The gel tank is designed for use with 8 cm x 10 cm mini gels and is compatible with gels cast on the mPAGE® Gel Caster and mPAGE® Precast Gels. The mPAGE® Mini Gel Tank is available in 2 formats for running up to 2 or 4 mini gels and is designed to prevent incorrect assembly and leakage.

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library