Skip to Content
Merck
All Photos(1)

Key Documents

CP45

Sigma-Aldrich

Anti-Clathrin Heavy Chain Mouse mAb (X22)

liquid, clone X22, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€501.00

€501.00


Estimated to ship on14 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€501.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€501.00


Estimated to ship on14 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

X22, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

bovine, mouse, rat, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLTCL1(8218)

General description

Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with Sp2/0-Ag14 mouse myeloma cells. Recognizes the ~180 kDa clathrin heavy chain protein.
Recognizes the ~180 kDa clathrin heavy chain protein.
This Anti-Clathrin Heavy Chain Mouse mAb (X22) is validated for use in Immunoblotting, Immunofluorescence, Immunoprecipitation for the detection of Clathrin Heavy Chain.

Immunogen

Human
clathrin heavy chain purified from human brain

Application

Immunoblotting (1:500)

Immunofluorescence (1:1000, indirect)

Immunoprecipitation (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Bauerfeind, R., et al. 1996. J. Neurocytol.25, 701.
Zhang, J.Z., et al. 1994. Cell78, 751.
Herskovits, J.S., et al. 1993. J. Cell Biol.122, 565.
Van Der Bliek, A.M., et al. 1993. J. Cell Biol.122, 553.
Pearse, B.M. and Robinson, M.S. 1990. Ann. Rev. Cell Biol.6, 151.
Brodsky, F.M. 1985. J. Cell Biol.101, 2047.
Immunoprecipitation of triskelions allows both the clathrin heavy chain and the associated light chain polypeptides to be examined. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dinah Loerke et al.
Traffic (Copenhagen, Denmark), 6(10), 918-929 (2005-09-06)
The clathrin triskelion is composed of three light chain (LC) and three heavy chain (HC) subunits. Cellular control of clathrin function is thought to be aimed at the LC subunit, mainly on the basis of structural information. To test this
Anna V Salova et al.
Oncotarget, 7(5), 6029-6047 (2015-12-31)
EGF complexed to fluorescent photostable quantum dots by biotin-streptavidin system (bEGF-savQD) is attractive for both the basic research and therapeutic application such as targeted drug delivery in EGF-receptor (EGFR) expressing cancers. However, compared to native EGF, the large size of
Vandana Kumari et al.
Journal of cystic fibrosis : official journal of the European Cystic Fibrosis Society, 16(3), 327-334 (2017-04-26)
AP2 is a clathrin-based endocytic adaptor complex comprising α, β2, μ2 and σ2 subunits. μ2 regulates CFTR endocytosis. The α subunit interacts with CFTR in the intestine but its physiologic significance is unclear. CFTR short circuit current was measured in
Stephanie A Zlatic et al.
Molecular biology of the cell, 24(15), 2378-2388 (2013-06-14)
A role for clathrin in AP-3-dependent vesicle biogenesis has been inferred from biochemical interactions and colocalization between this adaptor and clathrin. The functionality of these molecular associations, however, is controversial. We comprehensively explore the role of clathrin in AP-3-dependent vesicle
April Thelemann et al.
Molecular & cellular proteomics : MCP, 4(4), 356-376 (2005-01-20)
Overexpression and enhanced activation of the epidermal growth factor (EGF) receptor are frequent events in human cancers that correlate with poor prognosis. Anti-phosphotyrosine and anti-EGFr affinity chromatography, isotope-coded muLC-MS/MS, and immunoblot methods were combined to describe and measure signaling networks

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service