Skip to Content
Merck
All Photos(2)

Documents

CP2ALLRES

Milli-Q

Q-Gard® Purification Cartridge

Polishing cartridge for AFS® 8/16 and AFS® 8D/16D systems

Synonym(s):

Q-Gard® A Cartridge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
JA.14

Quality Level

packaging

pkg of 1 unit

manufacturer/tradename

Q-Gard®

technique(s)

ion chromatography: suitable

compatibility

for use with AFS®

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

The Q-Gard® A Cartridge is a polishing cartridge with virgin ion-exchange resins for use with AFS® 8/16 and AFS® 8D/16D water purification systems. This cartridge is designed to produce high resistivity water using carefully selected purification media.

Application

Q-Gard® A Cartridge is designed to remove ionic contaminants below trace level to match the water quality required for your application.

Features and Benefits

  • Guarantee water quality.
  • Ensure compliance with system specifications.
  • Employs ion-exchange technology for water purification.

Components

Virgin ion-exchange resins for ion removal

Legal Information

AFS is a registered trademark of Merck KGaA, Darmstadt, Germany
Q-GARD is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service