Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Check Cart for Availability | € 552,00 |
1 kit | Check Cart for Availability | € 963,00 |
About This Item
€ 552,00
usage
kit sufficient for ≤300 mL blood derivatives (CFMAG-100ML), kit sufficient for ≤750 mL blood derivatives (CFMAG-250ML)
technique(s)
DNA purification: suitable
input
plasma
serum
test parameters
: <45 min kit run time, sample volume: 0.1-10 mL
General description
Application
All
Preparation Note
Other Notes
Wash Buffer
Elution Buffer
Magnetic Bead solution
Legal Information
Disclaimer
1 of 1
This Item | ||
|---|---|---|
| usage kit sufficient for ≤300 mL blood derivatives (CFMAG-100ML), kit sufficient for ≤750 mL blood derivatives (CFMAG-250ML) | usage sufficient for 350 purifications | usage sufficient for 70 purifications |
| input plasma | input - | input blood |
| test parameters : <45 min kit run time, sample volume: 0.1-10 mL | test parameters - | test parameters : <40 min kit run time, sample volume: 200 μL |
| technique(s) DNA purification: suitable | technique(s) DNA purification: suitable | technique(s) DNA purification: suitable |
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1C
supp_hazards
Storage Class
8A - Combustible corrosive hazardous materials
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




