Skip to Content
Merck

Skip To

CDEX01201

Organex-Q® Cartridge

For Alpha-Q and Milli-Q® 3 / 4 Bowl systems

Synonym(s):

Organex-Q® Cartridge 30 cm (12")

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41104212
NACRES:
JA.14
eCl@ss:
33050190
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

feature

removes trace organics from water pretreated by reverse osmosis, deionization or distillation

packaging

pkg of 1 unit

L

30 cm (12 in.)

compatibility

for use with Alpha-Q (Before 1987), for use with Milli-Q® Bowl 3 system, for use with Milli-Q® Bowl 4 system

shipped in

ambient

General description

Organex-Q® Cartridge 30 cm (12") is a replacement cartridge used to remove trace organics from water pre-treated by reverse osmosis (RO), deionization or distillation. It is constructed using pure polypropylene and is equipped with thermally bonded endcaps to reduce the risk of extractables contamination.

Application

For use with Milli-Q® and Milli-RO bowl-style systems.

Features and Benefits

Purification method involves the use of ion-exchange and activated carbon.

Legal Information

MILLI-RO is a trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
ORGANEX-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CPEX02201CDFC01204QGARDTL01
description

For Alpha-Q and Milli-Q® 3 / 4 Bowl systems

description

Removes trace ionic and organic contaminants. For Super-Q® ultrapure high-flow water systems.

description

For Alpha-Q systems.

description

For Elix® Gulfstream/Large and RiOs Large systems connected to a pure water feed source.

compatibility

for use with Alpha-Q (Before 1987), for use with Milli-Q® Bowl 4 system, for use with Milli-Q® Bowl 3 system

compatibility

for use with Super-Q® Ultrapure High Flow Water Systems

compatibility

for use with Alpha-Q

compatibility

for use with Elix®, for use with Elix® Gulfstream, for use with Elix® Large, for use with RiOs Large

feature

removes trace organics from water pretreated by reverse osmosis, deionization or distillation

feature

removal of Inorganic and organic

feature

removes organics from water pretreated by reverse osmosis, deionization or distillation

feature

-

packaging

pkg of 1 unit

packaging

pkg of 1 unit

packaging

pkg of 4 units

packaging

pkg of 1 unit

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

L

30 cm (12 in.)

L

55 cm (22 in.)

L

30 cm (12 in.)

L

-

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300


Still not finding the right product?

Explore all of our products under Organex-Q® Cartridge




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–10 of 10 Questions  
  1. Este sistema tiene medidor de los diferentes parámetros? Magnesio, Calcio, PPM, Ph, Cloruro sódico, etc Qué capacidad diária tiene?

    1 answer
    1. The mentioned part number, CDEX01201, is a 12-inch Organex-Q cartridge typically used in 4-bowl systems (from a very long time ago).
      This cartridge has no fixed capacity in terms of liters of water throughput because it depends on the ionic content of the water as well as the dissolved carbon dioxide level. This cartridge has no meter directly, instead the water system in which it is installed has an analog resistivity meter.
      Best regards

      Helpful?

  2. What is the shelf life for Polisher?

    1 answer
    1. The shelf life is 2 years for all consumables kept in the recommended storage conditions (temperatures from 5 to 30°C; humidity between 10 and 80%). Contact your local representative (https://www.sigmaaldrich.com/collections/offices) for more information.

      Helpful?

  3. How are consumables changed?

    1 answer
    1. To help you to change your system's consumables, several video tutorials are available on our YouTube channel: https://youtube.com/playlist?list=PL2TUriaAaEA_IXDCnPbDk6OmrwptjwOHD. For high-flow systems, please keep in mind that all consumable changes must be approved by a 3-digit code that only the person in charge of the system knows.

      Helpful?

  4. How can I know which consumables can be used with my system?

    1 answer
    1. To know which consumables can be used on your system, you can see in the user manual of your system or see all the consumables for each system listed on https://www.sigmaaldrich.com/products/water-purification/milli-q-consumables.

      Helpful?

  5. What steps are you taking towards improved sustainability?

    1 answer
    1. Sustainability is at the heart of our innovations. From reducing water and energy consumption, to minimizing plastic and chemical waste, our dedicated water system engineers are constantly working on new ways to reduce the ecological footprint of our products to better support the sustainability goals we're all striving towards. For more information, we invite you to review the following brochure and infographic which highlight our efforts and achievements: https://www.sigmaaldrich.com/life-science/milliq#sus. Do not hesitate to contact your local representative or sales contact (https://www.sigmaaldrich.com/collections/offices) for further information.

      Helpful?

  6. Can I arrange to have my consumables automatically ordered and delivered once a year?

    1 answer
    1. It's possible to place only one consumable order per year with a specific contract called "Consumable Supply Agreement (CSA)" (https://www.sigmaaldrich.com/US/en/products/water-purification/milli-q-consumables#csa). For further information, contact your local representative or sales contact (https://www.sigmaaldrich.com/US/en/collections/offices).

      Helpful?

  7. How frequently do the various consumables need to be changed?

    1 answer
    1. The frequency that consumables must be changed depends on the system's usage (treated water volume) and on the consumables themselves (time of usage). In order to ensure the quality of water and your results, the consumables should be replaced when the system alerts you.

      Helpful?

  8. Can consumables be opened and refilled?

    1 answer
    1. Unfortunately, this is not possible because of safety issues and the need to guarantee the quality of consumables and their interior purification components. Our consumables are manufactured in an ISO 9001-certified facility France in agreement with high quality standards.

      Helpful?

  9. Where can I find data sheets, Certificates of Origin, Certificates of Analysis, and Certificates of Quality?

    1 answer
    1. Documents and certificates are available in the "Documentation" section on this page. Do not hesitate to contact us if you require further information.

      Helpful?

  10. What is the consumables delivery time?

    1 answer
    1. Refer to the consumable's Product page for a first estimation of its expected delivery time and contact customer service for more information.

      Helpful?

Reviews

No rating value

Active Filters