Skip to Content
Merck
All Photos(1)

Key Documents

CC314

Sigma-Aldrich

CytoSoft® Imaging 96-Well Plate

Elastic Moduli 0.5 kPa

Synonym(s):

96-Well Imaging Plate, CytoSoft Plate, Imaging 96-Well Plate

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€237.00

€237.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 EACH
€237.00

About This Item

UNSPSC Code:
12352207
NACRES:
NB.15

€237.00


Estimated to ship on28 May 2025


Request a Bulk Order

General description

The rigidity of the cell′s substrate (such as the extracellular matrix) can have a profound effect on cell function including morphology, differentiation and gene expression. Young′s elastic modulus is a mechanical property that measures the stiffness of a solid material. It defines the relationship between stress and strain in a material and is measured in kPa.

CytoSoft® plates provide a tool to culture cells on substrates with defined rigidities covering a broad physiological range (0.2 kPa- 64 kPa). On the bottom of each well, there is a thin layer of specially formulated biocompatible silicone, whose elastic modulus (rigidity) is carefully measured and certified. The surfaces of the gels are functionalized to form covalent bonds with amines on proteins. The gel readily binds to purified ECM proteins, such as PureCol type I collagen (5006), prior to cell addition.

CytoSoft® Imaging plates are designed for high-resolution imaging where low autofluorescence and exceptional optical clarity are required. These plates accommodate both live cell imaging and common fixation techniques such as immunostaining. Plate consists of a 175 μm thin polycarbonate film-bottom plate bonded with a black polystyrene frame and includes a lid. The plates are sterilized using ozone and provided with 1 plate per package.

Note: Use these recommendations as guidelines to determine the optimal coating conditions for your culture system. The bottom of the plate can be detached by excessive mechanical force such as centrifugation or direct contact with liquid handling tools (tips, pipettes, etc.).

1. Remove the CytoSoft® product from the protective sleeve in a sterile hood.
2. Prepare extracellular matrix material by neutralizing in amine-free buffer pH 7.4 to 7.9 (such as 1X DPBS). We do not recommend using gelatin as your ECM protein. Note: Pre-warm the coating solution to approximately room temperature before use
3. Dilute as needed, and dispense 150 μL of solution into each well to coat the surface. Note: Recommended dilution for PureCol Type I collagen is 1:30 (~100 μg/ml). Note: The hydrophobic surface requires larger volumes to cover the surface than do conventional plastic dishes
4. Incubate ECM coated CytoSoft® at room temperature, covered for 1 hour.
5. After incubation, aspirate any remaining material and rinse coated surfaces immediately two times with culture medium or PBS. Leave about 150 μL of medium per well to keep surface covered. Note: Do not allow the CytoSoft® surface to become dry once the surface has been wetted.
6. Coated surfaces are ready for use. Standard harvesting procedures used for removing cells from cultureware can be employed for harvesting cells from the CytoSoft® product including use of trypsin, Accutase and non-enzymatic cell detachment solutions.

CytoSoft® is a registered trademark of Advanced BioMatrix, Inc.

Storage and Stability

Store CytoSoft Imaging plates at room temperature.

Legal Information

CytoSoft is a registered trademark of Advanced BioMatrix, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

CytoSoft® modulus plates analyze substrate stiffness effects on cell behavior across a physiological range for mechanobiology research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service