Skip to Content
Merck

Skip To

CC086

Bovine Collagen Type VI

from bovine placenta villi, liquid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75
Biological source:
bovine
Form:
solution
Technique(s):
cell culture | mammalian: suitable
Concentration:
0.5 mg/mL
Assay:
>90% (SDS-PAGE)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Bovine Collagen Type VI, 0.5 mg/mL, solution, BioReagent, suitable for cell culture

biological source

bovine

Quality Segment

assay

>90% (SDS-PAGE)

form

solution

manufacturer/tradename

Chemicon®

concentration

0.5 mg/mL

technique(s)

cell culture | mammalian: suitable

input

sample type epithelial cells
sample type mesenchymal stem cell(s)
sample type hematopoietic stem cell(s)
sample type neural stem cell(s)
sample type pancreatic stem cell(s)
sample type: human embryonic stem cell(s)
sample type: mouse embryonic stem cell(s)

solubility

water: soluble at 20 °C

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

General description

Product Source: Bovine placenta villi

Application

Coating material for cell culture studies

Formation of collagen gels

Type VI collagen standard

Antigen for antibody production

Physical form

Liquid in 500 mM acetic acid, no preservatives.

Preparation Note

Collagen is extracted by controlled and limited pepsin digestion, followed by selective salt precipitation.
Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze thaws. The collagen may be transferred into physiological buffers by dialysis at 2-8°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CC077CC076CC054
biological source

bovine

biological source

human

biological source

human

biological source

human

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

assay

>90% (SDS-PAGE)

assay

95% (SDS-PAGE)

assay

95% (Human collagen type IV, SDS-PAGE)

assay

90% (Human collagen type III, SDS-PAGE)

concentration

0.5 mg/mL

concentration

1 mg/mL

concentration

1 mg/mL

concentration

1 mg/mL

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

form

solution

form

solution

form

liquid, solution

form

solution


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters