Skip to Content
Merck
All Photos(1)

Key Documents

CBL57

Sigma-Aldrich

Anti-Adrenocorticotropic Hormone Antibody, NT, clone 57

clone 57, Chemicon®, from mouse

Synonym(s):

ACTH

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€779.00

€779.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€779.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€779.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

clone

57, monoclonal

species reactivity

human, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Specificity

This antibody is specific for Synacthen (1-24 ACTH) and has no cross reaction with CLIP (ACTH 17-39). The antibody has an affinity constant greater than 10E11 L/mol.

KNOWN SPECIES CROSS REACTIVITY: Positive with rat N-terminal ACTH peptide

Application

This Anti-Adrenocorticotropic Hormone Antibody, N-terminus, clone 57 is validated for use in ELISA, IH(P) for the detection of Adrenocorticotropic Hormone.
This antibody reacts with solid phase ACTH either captured as part of a sandwich or adsorbed as a solid phase antigen and as such is suitable for serum assay development

The antibody is also reactive on formalin fixed paraffin wax embedded tissue and is therefore suitable for immunocyto-chemistry

Optimal working dilutions must be determined by the end user.

Linkage

Replaces: CBL58

Physical form

Format: Purified
The monoclonal is presented as 1mg in 1ml phosphate buffered saline containing 10mM sodium azide. We recommend that each laboratory determine an optimum working titre for use in its particular application.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service