Skip to Content
Merck
All Photos(1)

Key Documents

CBL468

Sigma-Aldrich

Anti-PECAM-1 Antibody, domains 3-6 of human PECAM-1, clone HC1/6

clone HC1/6, Chemicon®, from mouse

Synonym(s):

CD31

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€432.00

€432.00


Estimated to ship on23 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1216

Request a Bulk Order

Select a Size

Change View
100 μG
€432.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€432.00


Estimated to ship on23 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1216

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

HC1/6, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PECAM1(5175)

Specificity

Reacts with a 130-140 kDa transmembranous glycoprotein that is expressed widely amongst cells of the vascular compartment. CD31 is expressed on platelets, cultured endothelial cells, monocytes, normal tissue macrophages in tonsil, lung and kidney as well as in skin biopsies from conditions such as sarcoidosis and lepromatous leprosy.
FUSION PARTNER: P3-x63Ag8.653 myeloma cell line

Immunogen

Epitope: domains 3-6 of human PECAM-1

Application

Anti-PECAM-1 Antibody, domains 3-6 of human PECAM-1, clone HC1/6 is an antibody against PECAM-1 for use in FC, IH, IH(P) & IP.
Studies of the role of CD31 in leucocyte migration into inflammatory sites and in the process of angiogenesis

This antibody will stain frozen tissue sections

Suitable for immunoprecipitation studies

Suitable for flow cytometry either indirectly or directly when labelled with FITC or R-PE

Suitable for use on formalin fixed paraffin embedded sections after protease pre-treatment.

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified

Analysis Note

Control
Activated microglial cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fumiko Obata et al.
Toxins, 14(2) (2022-02-25)
Shiga toxin-producing Escherichia coli (STEC) causes proximal tubular defects in the kidney. However, factors altered by Shiga toxin (Stx) within the proximal tubules are yet to be shown. We determined Stx receptor Gb3 in murine and human kidneys and confirmed
Yu-Ting Huang et al.
Cellular signalling, 20(8), 1521-1527 (2008-05-27)
Sphingosine 1-phosphate (S1P) is a multifunctional phospholipid which acts through a specific family of G protein-coupled receptors. Platelet/endothelial cell adhesion molecule-1 (PECAM-1) form trans-homophilic binding at lateral cell border. Upon stimulation, its cytoplasmic tyrosine residues could be phosphorylated and interact
Guiting Lin et al.
Stem cells and development, 17(6), 1053-1063 (2008-07-04)
Adipose tissue-derived stem cells (ADSC) are routinely isolated from the stromal vascular fraction (SVF) of homogenized adipose tissue. Freshly isolated ADSC display surface markers that differ from those of cultured ADSC, but both cell preparations are capable of multipotential differentiation.
Gina Lisignoli et al.
Journal of cellular physiology, 220(2), 401-409 (2009-04-15)
Bone marrow stromal cells (MSCs) and osteoblasts are the two main non-haematopoietic cellular components of human bone tissue. To identify novel osteoblast-related molecules, we performed a gene expression profiling analysis comparing MSCs and osteoblasts isolated from the same donors. Genes
T M Herndon et al.
Journal of immunology (Baltimore, Md. : 1950), 166(9), 5654-5664 (2001-04-21)
The transcription factor NF-kappaB is a critical regulator of T cell function that becomes strongly activated in response to coengagement of TCR and CD28. Although events immediately proximal to NF-kappaB activation are well understood, uncertainty remains over which upstream signaling

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service