Skip to Content
Merck
All Photos(1)

Key Documents

CBL458

Sigma-Aldrich

Anti-CD49f Antibody, clone 4F10

clone 4F10, Chemicon®, from mouse

Synonym(s):

Integrin alpha6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€404.00

€404.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€404.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€404.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

4F10, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA6(3655)

Specificity

This antibody is specific to the integrin alpha6 chain which is expressed on epithelial and endothelial basement membrane. The VLA alpha6 chain, which consists of a single disulphide linked 120 kDa subunit and a small doublet subunit of 25/30 kDa, is non covalently associated with the VLA beta-chain (CD29) in VLA-6 or alpha6/beta1 complex or with integrin beta4 chain (alpha6/beta4).

The antigen is also expressed on platelets, monocytes, thymocytes and T lymphocytes.

FUSION PARTNER: NS1 myeloma cell line

Immunogen

Cell line SW/222 (Human colorectal carcinoma)

Application

Immunohistological studies on frozen sections

Immunoprecipitation of the laminin receptor

Flow Cytometry

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
Integrins
This Anti-CD49f Antibody, clone 4F10 is validated for use in FC, IP, IH for the detection of CD49f.

Linkage

Replaces: MAB2254

Physical form

Format: Purified
The monoclonal was purified by protein A affinity chromatography and is presented as a liquid in phosphate buffered saline containing 10mM sodium azide and 1 mg/mL bovine serum albumin.

Storage and Stability

Store an 2-8°C for up to 12 months from date of receipt. For long term storage aliquot antibody into small volumes and store at -20°C. Avoid repeated freeze-thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam Wahlsten et al.
Biomaterials, 273, 120779-120779 (2021-05-02)
The clinical treatment of large, full-thickness skin injuries with tissue-engineered autologous dermo-epidermal skin substitutes is an emerging alternative to split-thickness skin grafting. However, their production requires about one month of in vitro cell and tissue culture, which is a significant
Bryan W Day et al.
Cancer cell, 23(2), 238-248 (2013-02-16)
Significant endeavor has been applied to identify functional therapeutic targets in glioblastoma (GBM) to halt the growth of this aggressive cancer. We show that the receptor tyrosine kinase EphA3 is frequently overexpressed in GBM and, in particular, in the most
Laminin-5 induces osteogenic gene expression in human mesenchymal stem cells through an ERK-dependent pathway.
Klees, RF; Salasznyk, RM; Kingsley, K; Williams, WA; Boskey, A; Plopper, GE
Molecular Biology of the Cell null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service