Skip to Content
Merck

Skip To

CBL412A5

Anti-Tubulin β-III Antibody, clone TU-20, Alexa Fluor555 Conjugate

clone TU-20, from mouse, ALEXA FLUOR 555

Synonym(s):

Tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Check Cart for Availability
€ 545,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
ALEXA FLUOR 555
Clone:
TU-20, monoclonal
Application:
ICC
Citations:
2

€ 545,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

ALEXA FLUOR 555

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

TU-20, monoclonal

species reactivity

rat, pig

species reactivity (predicted by homology)

primate (based on 100% sequence homology), bovine (based on 100% sequence homology), mouse (based on 100% sequence homology), porcine (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Tubb3(22152)

General description

The unconjugated parent antibody (Cat. No. MAB1637) has an observed molecular weight at ~55 kDa
Tubulin is one of several members of a small family of globular proteins which polymerize to form cytoskeletal structures called microtubules. The most common members of the tubulin family are α-tubulin and β-tubulin. Tubulin dimers, typically composed of α- and β-tubulin subunits, assemble onto microtubules forming large polymers. β-III tubulin is a unique microtubule subunit that is found expressed almost exclusively in neurons. Due to the unique expression pattern of β-III tubulin it is often utilized as a marker to identify neuronal cells from other brain cells such as glia which do not express it. β-III tubulin also serves as a cytoarchitectural marker since it is primarily found within neurite extensions.

Immunogen

Epitope: C-terminus
Synthetic peptide corresponding to human Tubulin β-III.

Application

Detect Tubulin β-III using this Anti-Tubulin β-III Antibody, clone TU-20, Alexa Fluor555 Conjugate validated for use in ICC.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Biochem/physiol Actions

This antibody reacts with the C-terminus of the beta-III isoform of tubulin, which is known to be neuron specific. Does not identify beta tubulin found in glial cells.

Physical form

Protein G Purified
Purified mouse monoclonal IgG1 conjugated to Alexa Fluor 555 in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Preparation Note

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Rat E18 primary cortex cells
Evaluated by Immunocytochemistry in rat E18 primary cortex cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Tubulin β-III in rat E18 primary cortex cells.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T8328T5293T5201
clone

TU-20, monoclonal

clone

AA2, monoclonal

clone

2-28-33, monoclonal

clone

TUB 2.1, monoclonal

conjugate

ALEXA FLUOR 555

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Gene Information

mouse ... Tubb3(22152)

Gene Information

human ... TUBB(203068)

Gene Information

human ... TUBB1(81027), TUBB2A(7280), TUBB2C(10383)
rat ... Tubb2(29212), Tubb2c(296554)

Gene Information

human ... TUBB(203068), TUBB1(81027), TUBB1(81027), TUBB1(81027), TUBB1(81027), TUBB2A(7280), TUBB2A(7280), TUBB2A(7280), TUBB2A(7280), TUBB2C(10383), TUBB2C(10383), TUBB2C(10383), TUBB2C(10383)
mouse ... Tubb1(104068), Tubb1(104068), Tubb1(104068), Tubb1(104068), Tubb2a(22151), Tubb2a(22151), Tubb2a(22151), Tubb2a(22151), Tubb2c(227613), Tubb2c(227613), Tubb2c(227613), Tubb2c(227613), Tubb3(22152)
rat ... Tubb2(29212), Tubb2(29212), Tubb2(29212), Tubb2(29212), Tubb2c(296554), Tubb2c(296554), Tubb2c(296554), Tubb2c(296554)

species reactivity

rat, pig

species reactivity

human, mouse, rat, bovine

species reactivity

Ciona intestinalis, rat, chicken, Caenorhabditis elegans, Strongylocentrotus purpuratus, bovine, human

species reactivity

human, rat, frog, moth, mouse, plant, rabbit, chicken, bovine, wheat, sea urchin, hamster

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

antibody form

purified immunoglobulin

antibody form

purified from hybridoma cell culture

antibody form

ascites fluid

antibody form

purified immunoglobulin


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters