Skip to Content
Merck

Skip To

CBL309

Mouse Anti-Human IgG1 Antibody, clone 8c/6-39, Fc

clone 8c/6-39, Chemicon®, from mouse

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
0.5 mg
Check Cart for Availability
€ 666,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Clone:
8c/6-39, monoclonal
Species reactivity:
human
Application:
ELISA, IHC
Citations:
-

€ 666,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

8c/6-39, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable, immunohistochemistry: suitable

isotype

IgG2a

shipped in

wet ice

target post-translational modification

unmodified

Application

Immunohistochemical staining of acetone fixed tissue sections

ELISA assays

Mancini assays

Optimal working dilutions must be determined by the end user.
Mouse anti-Human IgG1 Antibody, clone 8c/6-39, Fc detects level of Human IgG1 & has been published & validated for use in ELISA, IH.

Biochem/physiol Actions

The antibody is specific for the CH2 domain of human IgG1 subclass

Physical form

Format: Purified
The monoclonal is presented in liquid in 0.01M phosphate buffered saline containing 0.1% sodium azide, pH7.1. We recommend that each laboratory determine an optimum working titre for use in its particular application.

Other Notes

Replaces: MABN1055

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B6775I2513SAB4200768
technique(s)

ELISA: suitable, immunohistochemistry: suitable

technique(s)

direct ELISA: 1:1,000

technique(s)

indirect ELISA: 1:400

technique(s)

ELISA: 1:8,000-1:16,000 using 5 μg/ml human IgG1 for coating

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

species reactivity

human

species reactivity

-

species reactivity

-

species reactivity

human

antibody form

affinity purified immunoglobulin

antibody form

purified from hybridoma cell culture

antibody form

ascites fluid

antibody form

purified from hybridoma cell culture

isotype

IgG2a

isotype

IgG2a

isotype

IgG2a

isotype

IgG2a

clone

8c/6-39, monoclonal

clone

8c/6-39, monoclonal

clone

8c/6-39, monoclonal

clone

8c/6-39, monoclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters