Skip to Content
Merck
All Photos(3)

Key Documents

CBL251

Sigma-Aldrich

Anti-Epithelial Specific Antigen Antibody, clone VU-1D9

clone VU-1D9, from mouse

Synonym(s):

Epithelial cell adhesion molecule, Ep-CAM, Adenocarcinoma-associated antigen, Cell surface glycoprotein Trop-1, Epithelial cell surface antigen, Epithelial glycoprotein, EGP, Epithelial glycoprotein 314, EGP314, hEGP314, KS 1/4 antigen, KSA, Major gastro

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€432.00

€432.00


Estimated to ship on25 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1215

Request a Bulk Order

Select a Size

Change View
100 μG
€432.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€432.00


Estimated to ship on25 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1215

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

VU-1D9, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

Epithelial specific antigen, also known as epithelial cell adhesion molecule (Ep-CAM) and epithelial glycoprotein (EGP), may act as an interaction molecule between intestinal epithelial cells (IECs) and intraepithelial lymphocytes (IELs) at the mucosal epithelium for providing immunological barrier as a first line of defense against mucosal infection. Epithelial specific antigen plays a role in embryonic stem cells proliferation and differentiation, and up-regulates the expression of FABP5, MYC, and cyclins A and E. Epithelial specific antigen is highly and selectively expressed by undifferentiated embryonic stem cells (ESCs). Protein levels rapidly diminish as soon as ESCs differentiate. Also expressed in most epithelial cell membranes and on the surface of adenocarcinoma.

Immunogen

NCI-H69 whole cells

Application

Anti-Epithelial Specific Antigen Antibody, clone VU-1D9 is a highly specific mouse monoclonal antibody, that targets Epithelial Cell Marker & has been tested in western blotting, ICC & IHC.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected Epithelial Specific Antigen in A431 cells.

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Epithelial Specific Antigen in human prostate adenocarcinoma cells.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

Evaluated by Western Blotting in MCF7 cell lyate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Epithelial Specific Antigen in 10 µg of MCF7 cell lyate.

Target description

~40 kDa observed. The calculated molecular weight of this protein is 35 kDa; however, this protein will run higher due to glycosylation.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
MCF7 cell lyasate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shin Young Park et al.
Oncology reports, 36(1), 197-204 (2016-05-14)
Regulated intramembrane proteolysis of epithelial cell adhesion molecule (EpCAM) results in shedding of the extracellular domain (EpEX) and release of the intra-cellular domain (EpICD) into the cytoplasm. Released EpICD associates with FHL2, β-catenin and Lef-1 to form a nuclear complex
Shaohua Wang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 36(6), e22336-e22336 (2022-05-07)
Idiopathic pulmonary fibrosis (IPF) and rheumatoid arthritis-associated interstitial lung disease (RA-ILD) are two fibrotic interstitial lung diseases that share the usual interstitial pneumonia (UIP) injury pattern. Here, we report that RNA sequencing of lung biopsies from patients with RA-ILD and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service