Skip to Content
Merck
All Photos(1)

Key Documents

CBL168F

Sigma-Aldrich

Anti-CD36 Antibody, clone SM-phi, FITC conjugated

clone SM-phi, Chemicon®, from mouse

Synonym(s):

Platelet Glycoprotein IV

Sign Into View Organizational & Contract Pricing

Select a Size

100 ASSAYS
€508.00

€508.00


Estimated to ship on10 May 2025


Request a Bulk Order

Select a Size

Change View
100 ASSAYS
€508.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

€508.00


Estimated to ship on10 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

SM-phi, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable

isotype

IgM

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD36(948)

Specificity

This antibody recognizes the 90 kDa single chain membrane glycoprotein (GPIIIb) that is found on monocytes/macrophages and platelets. FUSION PARTNER: NS1 myeloma cell line

Immunogen

Human tonsil cells and peripheral blood mononuclear cells

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
Studies of platelet and monocyte receptor for thrombospondin

Studies of platelet collagen adhesion receptor

Flow cytometry

Optimal working dilutions must be determined by the end user.
This Anti-CD36 Antibody, clone SM-phi, FITC conjugated is validated for use in FC, IH for the detection of CD36d. This antibody has been tested in Human.

Physical form

The monoclonal was purified by affinity chromatography and gel filtration prior to conjugation. The conjugate is supplied in 1.0 ml of phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. For flow cytometry we recommend using 10μL of conjugate per test.

Storage and Stability

Store at +4°C protected from light. DO NOT FREEZE. For long term use and storage aliquot conjugate into small volumes and store at +4°C for up to one year.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service