Skip to Content
Merck
All Photos(1)

Key Documents

BX1670

Supelco

2-Butanone

Meets ACS Specifications GR ACS

Synonym(s):

2-Butanone, Methyl Ethyl Ketone, Butanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-159-0
Assay:
≥99% (GC)
bp:
79.6 °C/1013 hPa
vapor pressure:
105 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

105 hPa ( 20 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

impurities

≤0.0005% Titrable acid
≤0.20% Water (H2O)

evapn. residue

≤0.0025%

color

APHA: ≤15

pH

7 (20 °C)

bp

79.6 °C/1013 hPa

mp

-86 °C

transition temp

flash point -4 °C

solubility

292 g/L

density

0.805 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

SMILES string

O=C(CC)C

InChI

1S/C4H8O/c1-3-4(2)5/h3H2,1-2H3

InChI key

ZWEHNKRNPOVVGH-UHFFFAOYSA-N

Application

Meets ACS Specifications GR ACS

Analysis Note

Assay (GC): 99.0% min
Color (APHA): 15 max
Evaporation residue: 0.0025% max
Form: Clear liquid
Infrared Spectrum: Conforms to standard
Titrable acid: 0.0005meq/g max
Water (H2O): 0.20% max

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

26.6 °F - closed cup

Flash Point(C)

-3 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service