Skip to Content
Merck
All Photos(1)

Key Documents

BM-114070

Millipore

Auto2D® IEF Chip pH4-7

used for first-dimension separations on the Auto2D® 2-D Electrophoresis Device.

Synonym(s):

2D Electrophoresis, 2DE, IEF, IEF Chip, IPG gel, precast gel

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€195.00

€195.00


Check Cart for Availability


Select a Size

Change View
1 PKG
€195.00

About This Item

UNSPSC Code:
41100000
NACRES:
NC.01

€195.00


Check Cart for Availability

General description

IEF Chip is 1st dimensional gel (separation medium) composed of IPG strip gel. IEF Chip pH4-7 is used for first-dimension separations on the Auto2D® 2-D Electrophoresis Device. This gives focused separation on pH4-7 range.

Application

  • Differential proteomic analysis
  • Post-translational modification
  • Protein Isoforms
  • Western Blotting

Legal Information

Auto2D is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service