Skip to Content
Merck
All Photos(1)

Documents

BM-10L-BK

Millipore

Anti-D (clones TH-28/MS-26) Blood Grouping Reagent

BIOSCOT Anti-D Blend from the clones TH-28/MS-26, Human IgM/IgG, Blood Grouping Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116102
NACRES:
NA.41

biological source

human

Quality Level

antibody form

unpurified

clone

TH-28 Human IgM/MS-26 Human IgG, monoclonal

form

liquid

packaging

pkg of 10 L

manufacturer/tradename

BIOSCOT

concentration

10-25 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

7.1-7.3

isotype

IgG
IgM

shipped in

ambient

storage temp.

2-8°C

General description

Appropriate for donor testing - reacts with all D weak and D partial red cells, including DVIContains 0.1% (w/v) Sodium AzideFiltered through a 0.22μm terminal filterShelf Life: 36 months from date of filtrationProduct formulation examined according to Annex IV of the EU IVD directive to support CE registration.For In Vitro use only.

Application

Bulk
For use by Tube;Slide;Microplate;Gel and Column techniques

Legal Information

BIOSCOT is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service