Skip to Content
Merck
All Photos(1)

Key Documents

BI0841

Sigma-Aldrich

Deblock Reagent 10% DCA in Toluene

Biosynthesis Novabiochem®

Synonym(s):

DCA Deblock, Deblock Reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H2Cl2O2
CAS Number:
Molecular Weight:
128.94
MDL number:
UNSPSC Code:
41116006
Pricing and availability is not currently available.

Quality Level

product line

Novabiochem®

form

liquid

reaction suitability

reaction type: Acetylations
reagent type: Lewis acid

transition temp

flash point 7.8 °C

shipped in

ambient

storage temp.

2-30°C

SMILES string

OC(=O)C(Cl)Cl

InChI

1S/C2H2Cl2O2/c3-1(4)2(5)6/h1H,(H,5,6)

InChI key

JXTHNDFMNIQAHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Biosynthesis Novabiochem®
DNA Reagents Label Change Explanation

Analysis Note

Color (APHA): 20APHA max
Dichloroacetic acid: 9.0-11.0%
Form: Clear liquid
Infrared Spectrum: Conforms
Water (H2O): 200ppm max

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Lact. - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A - STOT RE 2 Inhalation - STOT RE 2 Oral - STOT SE 3

Target Organs

Brain,Liver,Testes, Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

46.0 °F

Flash Point(C)

7.8 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the bottle cap screw size? Can we choose bottle cap screw size?

    1 answer
    1. The 4-liter bottle has a 38 mm closure. The 20 liters is packaged in a Nowpak container with a 63 mm closure. Unfortunately, the cap sizes are not customizable. However, the 20L package size has a bottle top adapter kit available for dispensing. Please see the link below to review this kit:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/148/300/011059-693001-ms.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service