Skip to Content
Merck
All Photos(1)

Key Documents

AP503

Sigma-Aldrich

Goat Anti-Mouse IgG γ chain Antibody, Species Adsorbed

1 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€209.00

€209.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€209.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

€209.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

goat

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

technique(s)

ELISA: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

General description

IgG is one of the most abundant protein in human serum. IgG is important for our defence against microorganisms and the molecules are produced by B-lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Specificity

Reacts with heavy chain of mouse IgG1, IgG2a, IgG2b and IgG3. Absorbed for mouse IgM and IgA, pooled human sera and purified human paraproteins. Minimal cross reactivity with human immunoglobulins.
The antibody reacts with the heavy chain of mouse IgG1, IgG2a, IgG2b and IgG3 as demonstrated by ELISA and flow cytometry. Minimal cross reactivity with human immunoglobulins.

Immunogen

Epitope: IgG γ chain
Prepared from purified mouse IgG γ chain. Pooled antisera from goats hyperimmunized with mouse IgG paraproteins.

Application

Goat anti-Mouse IgG γ chain Antibody, Species Adsorbed detects level of Mouse IgG γ chain & has been published & validated for use in ELISA.
Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling

Fragment Specific Secondary Antibodies

Physical form

Affinity Chromatography
Format: Purified
Purified by affinity chromatography on pooled mouse IgG covalently linked to agarose. Liquid in 1.0 mL of 100 mM borate buffered saline, pH 8.2.

Storage and Stability

Maintain refrigerated at 2°-8°C under sterile conditions for up to twelve months from date of receipt.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atsushi Kawai et al.
The Journal of clinical investigation, 133(23) (2023-12-01)
Intranasal vaccines are anticipated to be powerful tools for combating many infectious diseases, including SARS-CoV-2, because they induce not only systemic immunity but also mucosal immunity at the site of initial infection. However, they are generally inefficient in inducing an
Marcus Estrada et al.
Human vaccines & immunotherapeutics, 20(1), 2315709-2315709 (2024-02-19)
NDV-HXP-S is a Newcastle disease virus (NDV) vectored vaccine candidate which expresses the S-antigen of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2). This vaccine candidate is under evaluation in human clinical studies with and without cytosine phosphate guanine (CpG) 1018®
Eigo Kawahara et al.
Frontiers in immunology, 14, 1282016-1282016 (2024-01-04)
Respiratory syncytial virus (RSV) vaccines targeting the fusion glycoprotein (F protein) are highly effective clinically in preventing RSV challenges. The attachment glycoprotein (G protein) is a potentially effective vaccine antigen candidate, as it is important for cell adhesion during infection.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service