Skip to Content
Merck
All Photos(1)

Documents

AP502P

Sigma-Aldrich

Goat Anti-Human Ig κ chain Antibody, HRP conjugate, Species Adsorbed

Chemicon®, from goat

Synonym(s):

Anti-Human Ig HRP, HRP Goat Anti-Human Ig

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Antibody molecules typically comprise two immunoglobulin light chains covalently bound to a pair of heavy chains. Immunoglobulin light chains occur in two types, designated by the Greek letters kappa and lambda. Kappa and lambda light chains are approximately 250 amino acids in length with an average mass of about 25 kDa. The ratio of kappa to lambda found in the immunoglobulin population varies by species.

Specificity

Reacts with human kappa light chains. Absorbed for human myeloma proteins with λ light chains and mouse immunoglobulins.

Application

ELISA: 1:4,000-1:8,000.

Optimal working dilutions must be determined by the end user.
Goat anti-Human Ig κ chain Antibody, HRP conjugate, Species Adsorbed detects level of Human Ig κ chain & has been published & validated for use in ELISA.

Physical form

Purified by affinity chromatography on pooled human Igs with κ light chains covalently linked to agarose. Liquid 50% glycerol/50% PBS, pH 7.4.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyori Kim et al.
Oncology letters, 25(6), 236-236 (2023-05-08)
Anti-CD19 chimeric antigen receptor (CAR)-T cells have improved the outcomes of patients with B cell leukemia and lymphoma. However, their applications and positive outcomes remain limited. CAR-T cells are currently restricted to autologous blood as their source and their use
Ki Hyun Kim et al.
PLoS pathogens, 15(2), e1007375-e1007375 (2019-02-02)
Severe fever with thrombocytopenia syndrome (SFTS) is an emerging infectious disease localized to China, Japan, and Korea that is characterized by severe hemorrhage and a high fatality rate. Currently, no specific vaccine or treatment has been approved for this disease.
Ramin Dubey et al.
eLife, 9 (2020-05-21)
R-spondins (RSPOs) amplify WNT signaling during development and regenerative responses. We previously demonstrated that RSPOs 2 and 3 potentiate WNT/β-catenin signaling in cells lacking leucine-rich repeat-containing G-protein coupled receptors (LGRs) 4, 5 and 6 (Lebensohn and Rohatgi, 2018). We now
Jessica N Peel et al.
The Journal of clinical investigation, 134(8) (2024-03-12)
BACKGROUNDWeakly virulent environmental mycobacteria (EM) can cause severe disease in HLA-DRB1*15:02 or 16:02 adults harboring neutralizing anti-IFN-γ autoantibodies (nAIGAs). The overall prevalence of nAIGAs in the general population is unknown, as are the penetrance of nAIGAs in HLA-DRB1*15:02 or 16:02

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service