Sign In to View Organizational & Contract Pricing.
About This Item
UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Conjugate:
FITC conjugate
Clone:
polyclonal
Application:
IF
Citations:
1
biological source
rabbit
Quality Segment
conjugate
FITC conjugate
antibody form
affinity purified immunoglobulin
antibody product type
secondary antibodies
clone
polyclonal
species reactivity
sheep
manufacturer/tradename
Chemicon®
technique(s)
immunofluorescence: suitable
shipped in
wet ice
target post-translational modification
unmodified
Immunogen
Purified Sheep IgG.
Application
DILUTION:
Optimal working dilution must be determined by the end user.
Optimal working dilution must be determined by the end user.
Rabbit anti-Sheep IgG Antibody, FITC conjugate is an antibody against Sheep IgG for use in IF.
Research Category
Secondary & Control Antibodies
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies
Whole Immunoglobulin Secondary Antibodies
Biochem/physiol Actions
Sheep IgG (H&L)
Physical form
Lyophilized from 0.02M Phosphate buffer, 0.25 M NaCl with 15 mg/mL BSA and 0.1% sodium azide.
RECONSTITUTION:
Reconstitute to 1 mg/mL with sterile distilled water.
RECONSTITUTION:
Reconstitute to 1 mg/mL with sterile distilled water.
Preparation Note
Maintain lyophilized material at 2-8°C up to one year. After reconstitution maintain at 2-8°C in an undiluted format for up to 6 months.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Hazard Codes
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.