Skip to Content
Merck
All Photos(1)

Documents

AP1140

Sigma-Aldrich

Anti-GBA Mouse mAb (2E2)

liquid, clone 2E2, Calbiochem®

Synonym(s):

Anti-Glucosidase β, Acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2E2, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GBA(2629)

General description

Anti-GBA, mouse monoclonal, clone 2E2, recognizes the ~60 kDa GBA in MCF-7 and HeLa cells and human breast cancer tissue. It is validated for use in ELISA, WB, ICC, and IHC on paraffin sections.
Purified mouse monoclonal antibody. Recognizes the ~60 kDa GBA protein.
Recognizes the ~60 kDa GBA protein in MCF-7 and HeLa cells and human breast cancer tissue.

Immunogen

A recombinant polypeptide corresponding to amino acids of 146-236 human GBA, expressed as a GST fusion protein

Warning

Toxicity: Regulatory Review (Z)

Analysis Note

Negative Control
293T
Positive Control
MCF-7 cells, HeLa cells, Human breast cancer tissue

Other Notes

Campeau, P.M., et al. 2009. Blood114, 3181.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura J Smith et al.
Human molecular genetics, 32(5), 773-789 (2022-09-22)
Sequence variants or mutations in the GBA gene are numerically the most important risk factor for Parkinson disease (PD). The GBA gene encodes for the lysosomal hydrolase enzyme, glucocerebrosidase (GCase). GBA mutations often reduce GCase activity and lead to the
Ruggero Ferrazza et al.
Biochemical and biophysical research communications, 478(3), 1141-1146 (2016-08-20)
Mutations in LRRK2 gene cause inherited Parkinson's disease (PD) and variations around LRRK2 act as risk factor for disease. Similar to sporadic disease, LRRK2-linked cases show late onset and, typically, the presence of proteinaceous inclusions named Lewy bodies (LBs) in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service