Skip to Content
Merck
All Photos(1)

Key Documents

AP1135

Sigma-Aldrich

Anti-CTNNBL1 Mouse mAb (5F1)

liquid, clone 5F1, Calbiochem®

Synonym(s):

Anti-Beta catenin like protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€335.00

€335.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€335.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€335.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5F1, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG3

shipped in

ambient

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTNNBL1(56259)

General description

Purified mouse monoclonal antibody. Recognizes the ~65-70 kDa CTNNBL1 protein.
Recognizes the ~65-70 kDa CTNNBL1 protein in HeLa cells and human testes tissue.
This Anti-CTNNBL1 Mouse mAb (5F1) is validated for use in ELISA, Immunoblotting, Immunocytochemistry, Paraffin Sections for the detection of CTNNBL1.

Immunogen

A synthtic polypeptide corresponding to amino acids 210-311 of human rCTNNBL1
NA

Application




ELISA (see comments)
Immunoblotting (1-5 g/ml)
Immunocytochemistry (40 g/ml)
Paraffin Sections (3 g/ml)

Warning

Toxicity: Regulatory Review (Z)

Physical form

In PBS, pH 7.2

Reconstitution

Following initial, thaw aliquot and freeze (-20°C).

Analysis Note

Positive Control
HeLa cells, testes tissue

Other Notes

Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service