Skip to Content
Merck

Skip To

AG313

Glutamate Receptor δ 1/2 Control Peptide for AB1514

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.25
eCl@ss:
32160702
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


technique(s)

cell based assay: suitable

Quality Segment

UniProt accession no.

Gene Information

rat ... GRID1(79219)

General description

FUNCTION: SwissProt: Q9ULK0 # Receptor for glutamate. L-glutamate acts as an excitatory neurotransmitter at many synapses in the central nervous system. The postsynaptic actions of Glu are mediated by a variety of receptors that are named according to their selective agonists.
SIZE: 1009 amino acids; 112131 Da
SUBCELLULAR LOCATION: Membrane; Multi-pass membrane protein (By similarity).
SIMILARITY: SwissProt: Q9ULK0 ## Belongs to the glutamate-gated ion channel (TC 1.A.10) family.

Application

Antibody blocking (AB1514)

Optimal working concentration must be determined by the end user.

Physical form

Liquid

Preparation Note

Store at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library