Skip to Content
Merck

Skip To

AG243

Midkine, recombinant human

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352202
NACRES:
NA.41
eCl@ss:
32160405
Form:
powder
Assay:
>98% (SDS-PAGE and HPLC)
Biological source:
human


biological source

human

Quality Segment

assay

>98% (SDS-PAGE and HPLC)

form

powder

manufacturer/tradename

Chemicon®

impurities

<0.1 ng/mg Endotoxin (of MK)

UniProt accession no.

shipped in

dry ice

General description

Human Midkine (MK) is a new member of the heparin-binding neurotrophic factor family. MK and PTN (Pleiotrophin) are structural homologs, and are highly conserved among species. MK plays important roles in development and carcinogensis and has several important biological effects, including promotion of neurite extension and neuronal survival. Recombinant Midkine is a 13.4 kDa protein, comprising of 123 amino acid residues.
Product Source: E.coli

Physical form

Lyophilized from 0.4x PBS, pH 7.4.

Preparation Note

The lyophilized powder, though stable at room temperature, is best stored desiccated below 0°C. After a quick spin, reconstitute with sterile water to a concentration of 0.1-1.0 mg/mL. This solution can be diluted into water or other buffered solutions. Reconstitute MK should be stored in undiluted aliquots at -20°C for up to 6 months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

11 - Combustible Solids

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters