Skip to Content
Merck

Skip To

ACK5050PG

Amicon® Pro Affinity Concentration Kit Protein G with 50kDa Amicon® Ultra-0.5 Device

Amicon Pro Affinity Concentration Kit Protein G - A Centrifugal Tool for purifying & concentrating Antibodies.

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
41104916
NACRES:
NB.24
eCl@ss:
36100101
Material:
Amicon Ultra styrene-butadiene device, Ultracel® regenerated cellulose membrane (Amicon Ultra-0.5; low binding), polyethylene cap, polyethylene frit, polypropylene device (for exchange), polypropylene frit, polypropylene tube (AU0.5; for collection), polypropylene tube (for collection), polypropylene tube (holder)
Pore size:
50 kDa NMWCO
Working volume:
10 mL
Sterility:
non-sterile


material

Amicon Ultra styrene-butadiene device, Ultracel® regenerated cellulose membrane (Amicon Ultra-0.5; low binding), polyethylene cap, polyethylene frit, polypropylene device (for exchange), polypropylene frit, polypropylene tube (AU0.5; for collection), polypropylene tube (for collection), polypropylene tube (holder)

sterility

non-sterile

product line

Amicon®

manufacturer/tradename

Amicon® Pro

parameter

10 mL sample volume

technique(s)

buffer exchange: suitable, protein purification: suitable (antibody labeling), protein purification: suitable (concentration), protein purification: suitable (enrichment)

L

12 cm (4.7 in.)

diam.

2.97 cm

filtration area

1 cm2

working volume

10 mL

pore size

50 kDa NMWCO

shipped in

wet ice

General description

Filter Code: PLCGC
Filter Type: Ultrafiltration
This kit includes 12/pk Amicon Pro, 12/pk Amicon Ultra NMWCO 50 kDa, 3 ml Protein G resin (Binding capacity: 20mg human IgG/ml agarose), 2 X 66ml Bind/Wash Buffer, 12 ml Elution buffer, 2.5 ml Neutralization buffer

Features and Benefits

Minimum Final Concentrate Volume: 15 µL

Physical form

Color Code: Clear with pink cap

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2 - Met. Corr. 1

Storage Class

3 - Flammable liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters