Skip to Content
Merck

ACK5030PG

Amicon® Pro Affinity Concentration Kit Protein G with 30kDa Amicon® Ultra-0.5 Device

Amicon Pro Affinity Concentration Kit Protein G - A Centrifugal Tool for purifying & concentrating Antibodies.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41104916
NACRES:
NB.24
eCl@ss:
36100101
Material:
Amicon Ultra styrene-butadiene device, Ultracel® regenerated cellulose membrane (Amicon Ultra-0.5; low binding), polyethylene cap, polyethylene frit, polypropylene device (for exchange), polypropylene frit, polypropylene tube (AU0.5; for collection), polypropylene tube (for collection), polypropylene tube (holder)
Pore size:
30 kDa NMWCO
Working volume:
10 mL
Sterility:
non-sterile
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

Amicon Ultra styrene-butadiene device, Ultracel® regenerated cellulose membrane (Amicon Ultra-0.5; low binding), polyethylene cap, polyethylene frit, polypropylene device (for exchange), polypropylene frit, polypropylene tube (AU0.5; for collection), polypropylene tube (for collection), polypropylene tube (holder)

sterility

non-sterile

product line

Amicon®

manufacturer/tradename

Amicon® Pro

parameter

10 mL sample volume

technique(s)

buffer exchange: suitable, protein purification: suitable (antibody labeling), protein purification: suitable (concentration), protein purification: suitable (enrichment)

L

12 cm (4.7 in.)

diam.

2.97 cm

filtration area

1 cm2

working volume

10 mL

pore size

30 kDa NMWCO

shipped in

wet ice

General description

Filter Code: PLCGC
Filter Type: Ultrafiltration
This kit includes 12/pk Amicon Pro, 12/pk Amicon Ultra NMWCO 30 kDa, 3 ml Protein G resin Binding capacity: 20mg human IgG/ml agarose), 2 X 66ml Bind/Wash Buffer, 12 ml Elution buffer, 2.5 ml Neutralization buffer

Application

Amicon Pro Affinity Concentration Kit Protein G - A Centrifugal Tool for purifying & concentrating Antibodies.

Features and Benefits

Minimum Final Concentrate Volume: 15 µL

Physical form

Color Code: Clear with pink cap

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2 - Met. Corr. 1

Storage Class

3 - Flammable liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library