ABT80
Anti-Girdin, coiled-coil region Antibody
from rabbit, purified by affinity chromatography
Select a Size
About This Item
Recommended Products
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
monkey, human
species reactivity (predicted by homology)
rat
technique(s)
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
NCBI accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... CCDC88A(55704)
General description
Specificity
Immunogen
Application
Western Blot Analysis: A representative lot detected Girdin in Cos-7 cell lysate. Uncharacterized bands may be observed in some cell lysates.
Immunoprecipitation Analysis: A representative lot immunoprecipitated Girdin in HeLa cell lysates (Prof. P. Ghosh, Department of Medicine, Division of Gastroenterology and Hepatology, UCSD School of Medicine and Veterans Affairs Medical Center). Uncharacterized bands may be observed in some cell lysates.
Cell Structure
Cytoskeletal Signaling
Quality
Target description
Physical form
Storage and Stability
Analysis Note
HeLa cells
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service